AOBH3CD9F30A AOBChem

T-Butyl 4-bromo-1-naphthoate

  • MDL number MFCD09258636
  • Cas Number : 929000-22-6
  • Molecular Weight : 307.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/t-butyl-4-bromo-1-naphthoate.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/20482.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart