AOBH3CD9F311 AOBChem

2-(Morpholinomethyl)phenylboronic acid pinacol ester

  • MDL number MFCD00778120
  • Cas Number : 876316-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-Morpholinomethyl-phenylboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/21242.pdf
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart