AOBH3CD9F32D AOBChem

2-(3-Bromo-5-nitro-4-pyridinyl)-N,N-dimethylethenamine

  • MDL number MFCD05244519
  • Cas Number : 1258875-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-3-bromo-5-nitro-4-pyridinyl-n-n-dimethylethenamine.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/23147.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart