AOBH3CD9F32E AOBChem

Diethyl (3-bromo-5-nitropyridin-4-yl)malonate

  • Cas Number : 69872-14-6
  • Molecular Weight : 361.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/diethyl-3-bromo-5-nitropyridin-4-yl-malonate.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/23148.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart