AOBH3CD9F335 AOBChem

1,2-Difluoro-4,5-dimethoxybenzene

  • Cas Number : 203059-80-7
  • Molecular Weight : 174.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/1-2-difluoro-4-5-dimethoxybenzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/23421.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart