AOBH3CD9F431 AOBChem

2-(4-(4-Fluorophenoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 1355052-34-4
  • Molecular Weight : 314.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/2-4-4-fluorophenoxy-phenyl-4-4-5-5-tetramethyl-1-3-2-dioxaborolane.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/12725.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart