AOBH3CD9F43B AOBChem

5-(4-bromo-3-fluorophenyl)oxazole

  • Cas Number : 2111410-80-9
  • Molecular Weight : 242.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-4-bromo-3-fluorophenyl-oxazole.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/35983.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart