AOBH3CD9F44D AOBChem

4-(4-Methylpiperazine-1-carbonyl)phenylboronic acid hydrochloride

  • MDL number MFCD09027217
  • Cas Number : 913835-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/4-4-methylpiperazine-1-carbonyl-phenylboronic-acid-hydrochloride.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/29355.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart