AOBH3CD9F467 AOBChem

(2,3-difluorophenyl)(pyrrolidin-1-yl)methanone

  • Cas Number : 385380-62-1
  • Molecular Weight : 211.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-3-difluorophenyl-pyrrolidin-1-yl-methanone.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/36041.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart