AOBH7BADDAEE AOBChem

5-Bromo-2-(bromomethyl)pyridine hydrobromide

  • Cas Number : 173999-22-9
  • Molecular Weight : 331.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/5-bromo-2-bromomethyl-pyridine-hydrobromide.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/30243.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart