AOBH94550E9E AOBChem

3-Ethoxy-2-fluorobenzaldehyde

  • Cas Number : 1204176-29-3
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link https://aobchem.com/3-ethoxy-2-fluorobenzaldehyde.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/19876.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart