AOBH94550EA5 AOBChem

2-Propoxypyridine

  • Cas Number : 101870-22-8
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link https://aobchem.com/2-propoxypyridine.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/20062.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart