AOBH94550EAD AOBChem

3-Methylthioanisole

  • MDL number MFCD01683019
  • Cas Number : 4886-77-5
  • Molecular Weight : 138.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link https://aobchem.com/3-methylthioanisole.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/20285.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart