AOBH94550EDE AOBChem

2-Chloro-4-iodo-5-(methoxymethoxy)pyridine

  • Cas Number : 877133-57-8
  • Molecular Weight : 299.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link https://aobchem.com/2-Chloro-4-iodo-5-methoxymethoxy-pyridine.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/22595.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart