AOBH94550EEA AOBChem

1-Fluoro-4,5-dimethoxy-2-iodobenzene

  • Cas Number : 900175-06-6
  • Molecular Weight : 282.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link https://aobchem.com/1-fluoro-4-5-dimethoxy-2-iodobenzene.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/23001.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart