AOBH94550EEF AOBChem

(4-Bromo-phenyl)-piperidin-1-yl-methanone

  • Cas Number : 98612-93-2
  • Molecular Weight : 268.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link https://aobchem.com/4-bromo-phenyl-piperidin-1-yl-methanone.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/23217.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart