AOBH94550F1C AOBChem

3-Bromo-4-chloro-5-cyclopropylpyridine

  • Cas Number : 1404367-15-2
  • Molecular Weight : 232.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link https://aobchem.com/3-bromo-4-chloro-5-cyclopropylpyridine.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/26091.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart