AOBH94550F1F AOBChem

3-Bromo-2-fluorobenzamide

  • Cas Number : 871353-25-2
  • Molecular Weight : 218.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link https://aobchem.com/3-bromo-2-fluorobenzamide.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/26329.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart