AOBH94550F38 AOBChem

4-Iodo-2-(trifluoromethoxy)aniline

  • Cas Number : 874814-75-2
  • Molecular Weight : 303.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link https://aobchem.com/4-iodo-2-trifluoromethoxy-aniline.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/27714.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart