AOBH94550F49 AOBChem

1-[(4-Bromophenoxy)methyl]-3-fluorobenzene

  • Cas Number : 1019123-14-8
  • Molecular Weight : 281.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link https://aobchem.com/1-4-bromophenoxy-methyl-3-fluorobenzene.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/29540.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart