AOBH94551017 AOBChem

4-Fluoro-2-nitrophenylboronic acid

  • Cas Number : 860034-00-0
  • Molecular Weight : 184.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link https://aobchem.com/4-Fluoro-2-nitrophenylboronic-acid.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/13831.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart