AOBH9455103E AOBChem

5-Bromo-2-fluoro-4-methoxybenzaldehyde

  • Cas Number : 473417-48-0
  • Molecular Weight : 233.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link https://aobchem.com/5-bromo-2-fluoro-4-methoxybenzaldehyde.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/14858.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart