AOBH945510DB AOBChem

3-Chloro-5-(trifluoromethoxy)benzoic acid

  • Cas Number : 433926-46-6
  • Molecular Weight : 240.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link https://aobchem.com/3-chloro-5-trifluoromethoxy-benzoic-acid.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/15750.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart