AOBH9455117C AOBChem

5-Bromo-3-fluoro-2-(phenylmethoxy)benzaldehyde,

  • Cas Number : 1003298-75-6
  • Molecular Weight : 309.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link https://aobchem.com/5-bromo-3-fluoro-2-phenylmethoxy-benzaldehyde.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/17005.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart