AOBH945511ED AOBChem

4-Carboxy-3-isopropylphenylboronic acid

  • Cas Number : 851335-14-3
  • Molecular Weight : 208.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link https://aobchem.com/4-carboxy-3-isopropylphenylboronic-acid.html

Safety Information

MSDS https://aobchem.com/pub/media/files/sds/19500.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart