APO414278154 Apollo Scientific Ltd

Methyl 3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate

  • MDL number MFCD07774184
  • Cas Number : 142166-00-5
  • Molecular Weight : 207.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 247-253
Iupac Name methyl 3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate
Inchi Code 1S/C10H9NO4/c1-14-10(13)6-2-3-7-8(4-6)15-5-9(12)11-7/h2-4H,5H2,1H3,(H,11,12)
Inchi Key HBGXBRNLTSFTIG-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 247-253
Product Link https://store.apolloscientific.co.uk/product/methyl-3-oxo-34-dihydro-2h-14-benzoxazine-7-carboxylate

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/methyl-3-oxo-34-dihydro-2h-14-benzoxazine-7-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart