APO414278198 Apollo Scientific Ltd

5-Bromo-2-nitrobenzaldehyde

  • MDL number MFCD00456508
  • Cas Number : 20357-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-73
Synonym 4-Bromo-2-formylnitrobenzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-nitrobenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-nitrobenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart