APO414278239 Apollo Scientific Ltd

6-Bromo-1-cyclopentyl-1H,2H,3H-imidazo[4,5-b]pyridin-2-one

  • MDL number MFCD22383941
  • Cas Number : 1393845-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-168
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52820

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52820
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart