APO414278288 Apollo Scientific Ltd

2-(4-Bromophenyl)-5-methyl-1,3,4-oxadiazole

  • MDL number MFCD00491620
  • Cas Number : 41421-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-110
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-4-bromophenyl-5-methyl-134-oxadiazole

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-4-bromophenyl-5-methyl-134-oxadiazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart