APO414278294 Apollo Scientific Ltd

6-Chloro-N4-methylpyrimidine-2,4-diamine

  • MDL number MFCD02091108
  • Cas Number : 1005-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159-162
Synonym 2-Amino-4-chloro-6-(methylamino)pyrimidine, 2-Amino-4-chloro-6-(methylamino)-1,3-diazine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54341

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54341
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart