APO414278339 Apollo Scientific Ltd

3-Methyl-1-[2-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

  • MDL number MFCD02715834
  • Cas Number : 380238-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Amino-3-methyl-1-[2-(trifluoromethyl)phenyl]-1H-pyrazole, 2-(5-Amino-3-methyl-1H-pyrazol-1-yl)benzotrifluoride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-methyl-1-2-trifluoromethylphenyl-1h-pyrazol-5-amine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-methyl-1-2-trifluoromethylphenyl-1h-pyrazol-5-amine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart