APO414278379 Apollo Scientific Ltd

Ethyl 3,8-dichloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD17168289
  • Cas Number : 1355171-49-1
  • Molecular Weight : 327.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-38-dichloro-6-trifluoromethylimidazo12-apyridine-2-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-38-dichloro-6-trifluoromethylimidazo12-apyridine-2-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart