APO414278399 Apollo Scientific Ltd

Ethyl 3-amino-5-bromo-1-benzothiophene-2-carboxylate

  • MDL number MFCD14635904
  • Cas Number : 1308649-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53939

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53939
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart