APO414278664 Apollo Scientific Ltd

3-Acetyl-5-bromo-1H-indazole

  • MDL number MFCD07374362
  • Cas Number : 886363-74-2
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(5-Bromo-1H-indazol-3-yl)ethan-1-one, 3-Ethanoyl-5-bromo-1H-indazole
Purity 96
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-acetyl-5-bromo-1h-indazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-acetyl-5-bromo-1h-indazole
pictograms GHS07
hstatements H302+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart