APO414278725 Apollo Scientific Ltd

N-Methyl-2-(trifluoromethyl)benzenesulfonamide

  • MDL number MFCD05270097
  • Cas Number : 633697-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53544

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53544
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart