APO414278746 Apollo Scientific Ltd

4-(1H-Pyrazol-3-yl)pyridine

  • MDL number MFCD00115150
  • Cas Number : 17784-60-0
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157-158
Density 1.391
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-1h-pyrazol-3-ylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-1h-pyrazol-3-ylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart