APO414278749 Apollo Scientific Ltd

5-Chloro-2-(methylamino)nicotinonitrile

  • MDL number MFCD15146802
  • Cas Number : 1427460-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53976

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53976
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart