APO414278754 Apollo Scientific Ltd

Methyl 2-(chloromethyl)-1,3-benzoxazole-7-carboxylate

  • MDL number MFCD18804150
  • Cas Number : 1355170-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54057

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54057
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart