APO414278917 Apollo Scientific Ltd

8-Bromoimidazo[1,2-a]pyridine-6-carbonitrile

  • MDL number MFCD14584821
  • Cas Number : 1221791-93-0
  • Molecular Weight : 222.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-186
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-bromoimidazo12-apyridine-6-carbonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/8-bromoimidazo12-apyridine-6-carbonitrile
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart