APO414278921 Apollo Scientific Ltd

Ethyl 6,8-dibromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD22421937
  • Cas Number : 859787-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53796

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53796
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart