APO414278940 Apollo Scientific Ltd

Methyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate

  • MDL number MFCD08706234
  • Cas Number : 35212-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-3-amino-4-methoxy-1-benzothiophene-2-carboxylate

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/methyl-3-amino-4-methoxy-1-benzothiophene-2-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart