APO414278946 Apollo Scientific Ltd

1H-Indazole-4-carbohydrazide

  • MDL number MFCD11506090
  • Cas Number : 1086392-16-6
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1H-indazole-4-carbohydrazide
Inchi Code 1S/C8H8N4O/c9-11-8(13)5-2-1-3-7-6(5)4-10-12-7/h1-4H,9H2,(H,10,12)(H,11,13)
Inchi Key LIQJWNMFLQUVNT-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1h-indazole-4-carbohydrazide

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/1h-indazole-4-carbohydrazide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart