APO414278958 Apollo Scientific Ltd

3-Bromo-1H-indazol-7-amine

  • MDL number MFCD09027010
  • Cas Number : 316810-90-9
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 135
Iupac Name 3-bromo-1H-indazol-7-amine
Inchi Code 1S/C7H6BrN3/c8-7-4-2-1-3-5(9)6(4)10-11-7/h1-3H,9H2,(H,10,11)
Inchi Key GFCDWESCMQBURY-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 135
Product Link https://store.apolloscientific.co.uk/product/3-bromo-1h-indazol-7-amine

Safety Information

signalword Warning
pcodes P280;P301+P310
MSDS https://store.apolloscientific.co.uk/product/3-bromo-1h-indazol-7-amine
pictograms GHS07
hstatements H302;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart