APO414278983 Apollo Scientific Ltd

1H-Benzimidazole-5-carbohydrazide

  • MDL number MFCD08703627
  • Cas Number : 108038-52-4
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1H-benzimidazole-5-carbohydrazide
Inchi Code 1S/C8H8N4O/c9-12-8(13)5-1-2-6-7(3-5)11-4-10-6/h1-4H,9H2,(H,10,11)(H,12,13)
Inchi Key XMKCHQVJPWPSPQ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1h-benzimidazole-5-carbohydrazide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1h-benzimidazole-5-carbohydrazide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart