APO414279033 Apollo Scientific Ltd

Ethyl 3-bromoimidazo[1,2-a]pyridine-6-carboxylate

  • MDL number MFCD12827886
  • Cas Number : 1215504-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-bromoimidazo12-apyridine-6-carboxylate

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-bromoimidazo12-apyridine-6-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart