APO414279128 Apollo Scientific Ltd

Methyl 2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoate

  • MDL number MFCD25976394
  • Cas Number : 96017-05-9
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-1-oxo-13-dihydro-2h-isoindol-2-ylpropanoate

Safety Information

MSDS https://store.apolloscientific.co.uk/product/methyl-2-1-oxo-13-dihydro-2h-isoindol-2-ylpropanoate
Availability details for
Quantity
  • Availability
  • Add to Cart