APO414279129 Apollo Scientific Ltd

Methyl 3-methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoate

  • MDL number MFCD25976395
  • Cas Number : 1268618-92-3
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-methyl-2-1-oxo-13-dihydro-2h-isoindol-2-ylbutanoate

Safety Information

MSDS https://store.apolloscientific.co.uk/product/methyl-3-methyl-2-1-oxo-13-dihydro-2h-isoindol-2-ylbutanoate
Availability details for
Quantity
  • Availability
  • Add to Cart