APO414279181 Apollo Scientific Ltd

8-Bromo-1-hydroxyisoquinoline

  • MDL number MFCD12761600
  • Cas Number : 475994-60-6
  • Molecular Weight : 224.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Bromoisoquinolin-1-ol, 8-Bromo-1-hydroxy-2-azanaphthalene
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-bromo-1-hydroxyisoquinoline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/8-bromo-1-hydroxyisoquinoline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart