APO414279211 Apollo Scientific Ltd

3-Bromo-7-chloroimidazo[1,2-a]pyridine

  • MDL number MFCD11111834
  • Cas Number : 342613-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-7-chloroimidazo12-apyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-7-chloroimidazo12-apyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart