APO414279248 Apollo Scientific Ltd

4,4',4''-[(Benzene-1,3,5-triyl)triethyne-2,1-diyl]tripyridine

  • MDL number MFCD25541760
  • Cas Number : 168289-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237(dec.)
Synonym 1,3,5-Tris[(pyridin-4-yl)ethynyl]benzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54867

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54867
pictograms GHS07
hstatements H302+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart